MilliporeSigma
  • Molecular genetic analysis of the Jk(a-b-) phenotype in Chinese: A novel silent recessive JK allele.
Molecular genetic analysis of the Jk(a-b-) phenotype in Chinese: A novel silent recessive JK allele.
Transfusion and apheresis science : official journal of the World Apheresis Association : official journal of the European Society for Haemapheresis (2016-03-13)
Ai Zhang, Quan Chi, Hongkeng Lin, Yimin She
ABSTRACT
The Jk(a-b-) phenotype, referred to as Jknull, is rare in most populations. This blood type is characterized by the absence of Kidd glycoprotein on the surface of red blood cells (RBCs) and moderately reduced ability to concentrate urine. The molecular basis for Jknull phenotype includes splice-site mutations, missense mutations, and a partial gene deletion in the JK(SLC14A1) gene that encodes the human urea transporter protein. In this study, we have analyzed 10 Chinese Jknull samples to determine their molecular bases. In addition to the well known Polynesian Jknull allele, three Jknull alleles were detected including one novel Jknull allele: JKA (130A, 220G).