MilliporeSigma
  • Novel mutations of the RS1 gene in a cohort of Chinese families with X-linked retinoschisis.
Novel mutations of the RS1 gene in a cohort of Chinese families with X-linked retinoschisis.
Molecular vision (2014-02-08)
Jieqiong Chen, Ke Xu, Xiaohui Zhang, Zhe Pan, Bing Dong, Yang Li
ABSTRACT
X-linked retinoschisis is a retinal dystrophy caused by mutations in the RS1 gene in Xp22.1. These mutations lead to schisis (splitting) of the neural retina and subsequent reduction in visual acuity in affected men (OMIM # 312700). The aim of this study was to identify the RS1 gene mutations in a cohort of Chinese patients with X-linked retinoschisis, and to describe the associated phenotypes. Patients and unaffected individuals from 16 unrelated families underwent detailed ophthalmic examinations. After informed consent was obtained, genomic DNA was extracted from the venous blood of all participants. All exons including the exon-intron boundaries of the RS1 gene, were amplified by PCR and the products were analyzed by direct sequencing. Long-range PCR followed by DNA sequencing was used to define the breakpoints of the large deletion. Sixteen male individuals from 16 families were diagnosed with retinoschisis by clinical examination. The median age at review was 13.2 years (range: 5-34 years); the median best-corrected visual acuity upon review was 0.26 (range 0.02-1.0). Foveal schisis was found in 82.8% of the eyes (24/29) while peripheral schisis was present in 27.5% of the eyes (8/29). Sequencing of the RS1 gene identified 16 mutations, nine of which were novel. The mutations included eight missense mutations, all located in exons 4-6 (50.0%), two nonsense mutations (12.5%), four small deletions or insertions (25.0%), one splice site mutation (6.25%), and one large genomic deletion that included exon1 (6.25%). The mutations found in our study broaden the spectrum of RS1 mutations. The identification of the specific mutation in each pedigree will allow future determination of female carrier status for genetic counseling purposes.