MilliporeSigma
  • Proteomic analysis reveals O-GlcNAc modification on proteins with key regulatory functions in
Proteomic analysis reveals O-GlcNAc modification on proteins with key regulatory functions in
Proceedings of the National Academy of Sciences of the United States of America (2017-02-06)
Shou-Ling Xu, Robert J Chalkley, Jason C Maynard, Wenfei Wang, Weimin Ni, Xiaoyue Jiang, Kihye Shin, Ling Cheng, Dasha Savage, Andreas F R Hühmer, Alma L Burlingame, Zhi-Yong Wang
ABSTRACT
Genetic studies have shown essential functions of O-linked
Sigma-Aldrich
PUGNAc, ≥95% (HPLC)