MilliporeSigma
  • Suppression of HSP27 Restores Retinal Function and Protects Photoreceptors From Apoptosis in a Light-Induced Retinal Degeneration Animal Model.
Suppression of HSP27 Restores Retinal Function and Protects Photoreceptors From Apoptosis in a Light-Induced Retinal Degeneration Animal Model.
Investigative ophthalmology & visual science (2017-06-21)
Chih-Cheng Chien, Chi-Jung Huang, Lu-Tai Tien, Yu-Che Cheng, Chia-Ying Ke, Yih-Jing Lee
ABSTRACT
We used a light-induced retinal degeneration animal model to investigate possible roles of heat shock protein 27 (HSP27) in retinal/photoreceptor protection. Sprague-Dawley rats were used for the light-induced retinal degeneration animal model. The histology of eye sections was observed for morphologic changes in the retina. Cell apoptosis was examined in each group using the terminal deoxynucleotidyl transferase dUTP nick-end labeling assay, and electroretinography was used to evaluate retinal function. Protein and mRNA expression levels of different retinal cell markers were also detected through immunofluorescence staining, Western blotting, and real-time PCR. The thickness of the outer nuclear layer significantly decreased after 7-day light exposure. Moreover, we injected a viral vector for silencing HSP27 expression into the eyes and observed that photoreceptors were better preserved in the HSP27-suppressed (sHSP27) retina 2 weeks after injection. HSP27 suppression also reduced retinal cell apoptosis caused by light exposure. In addition, the loss of retinal function caused by light exposure was reversed on suppressing HSP27 expression. We subsequently found that the expression of the Rho gene and immunofluorescence staining of rhodopsin and arrestin (cell markers for photoreceptors) increased in sHSP27-treated retinas. HSP27 suppression did not affect the survival of ganglion and amacrine cells. Retinal cell apoptosis and functional loss were observed after 7-day light exposure. However, in the following 2 weeks after light exposure, HSP27 suppression may initiate a protective effect for retinal cells, particularly photoreceptors, from light-induced retinal degeneration.