MilliporeSigma
  • Neurochemical phenotypes of endomorphin-2-containing neurons in vagal nodose neurons of the adult rat.
Neurochemical phenotypes of endomorphin-2-containing neurons in vagal nodose neurons of the adult rat.
Neurochemistry international (2009-05-26)
Le Niu, Tao Chen, Ya-Yun Wang, Yun-Qing Li
ABSTRACT
It has been shown that endomorphin-2-like immunoreactive (EM2-LI) neurons in dorsal root ganglion play important roles in regulating somatic information transmission. Although EM2-ergic neurons have been found in nodose ganglion (NG) which is mainly involved in transmitting visceral information into the nucleus tractus solitarii (NTS), the neurochemical phenotypes of EM2-ergic neurons have not yet been investigated. In the present study, immunofluorescent histochemical staining showed that 43.5% of the NG neurons contained EM2 and these neurons were small to medium in size. 15.2%, 27.8%, 74.4% and 25.2% of the EM2-LI NG neurons expressed substance P (SP), calcitonin gene-related peptide (CGRP), nitric oxide synthase (NOS) and vasoactive intestinal peptide (VIP), respectively. In addition, about 90.8% of EM2-LI NG neurons also contained mu-opioid receptor (MOR). EM2/MOR and EM2/SP double-labeled peripheral axons were observed in the vagal trunk. Anterograde tracing combined with immunofluorescent staining showed EM2/MOR and EM2/SP double-labeled vagal afferents in the NTS. EM2/MOR/SP and EM2/MOR/CGRP triple-labeled neurons and axons were observed in the NG. Importantly, at the ultrastructrual level, post-embedding electron microscopy revealed that EM2-LI and SP-LI gold particles coexisted in the same large dense-cored synaptic vesicles in the pre-synaptic button, while MOR-LI gold particles existed on both pre- and post-synaptic membranes in the NTS. These results suggest that EM2 in axon terminals of NG neurons might be involved in visceral information transmission and homeostatic control through modulating the release of other neurotransmitters (such as SP, CGRP, NO, VIP) via pre-synaptic MOR and through post-synaptic mechanisms in the NTS.