MilliporeSigma
  • Treatment of mammary carcinomas in HER-2 transgenic mice through combination of genetic vaccine and an agonist of Toll-like receptor 9.
Treatment of mammary carcinomas in HER-2 transgenic mice through combination of genetic vaccine and an agonist of Toll-like receptor 9.
Clinical cancer research : an official journal of the American Association for Cancer Research (2009-02-26)
Luigi Aurisicchio, Daniela Peruzzi, Antonella Conforti, Sridhar Dharmapuri, Antonella Biondo, Saverio Giampaoli, Arthur Fridman, Ansu Bagchi, Christopher T Winkelmann, Raymond Gibson, Ekambar R Kandimalla, Sudhir Agrawal, Gennaro Ciliberto, Nicola La Monica
ABSTRACT
Oligodeoxynucleotides containing unmethylated CpG dinucleotides induce innate and adaptive immunity through Toll-like receptor 9 (TLR9). In the present study, we have examined the ability of a novel agonist of TLR9, called immunomodulatory oligonucleotide (IMO), to enhance effects of a HER-2/neu plasmid DNA electroporation/adenovirus (DNA-EP/Ad) vaccine. BALB/NeuT mice were treated with DNA-EP vaccine alone, IMO alone, or the combination of two agents starting at week 13, when all mice showed mammary neoplasia. Tumor growth and survival were documented. Antibody and CD8+ T-cell responses were determined. Peptide microarray analysis of sera was carried out to identify immunoreactive epitopes. Additionally, microCT and microPET imaging was carried out in an advanced-stage tumor model starting treatment at week 17 in BALB/NeuT mice. The combination of DNA-EP and IMO resulted in significant tumor regression or delay to tumor progression. 2-Deoxy-2-[18F]fluoro-D-glucose microPET and microCT imaging of mice showed reduced tumor size in the DNA-EP/IMO combination treatment group. Mice treated with the combination produced greater antibody titers with IgG2a isotype switch and antibody-dependent cellular cytotoxicity activity than did mice treated with DNA-EP vaccine. An immunogenic B-cell linear epitope, r70, within the HER-2 dimerization domain was identified through microarray analysis. Heterologous DNA-EP/Ad vaccination combined with IMO increased mice survival. The combination of HER-2/neu genetic vaccine and novel agonist of TLR9 had potent antitumor activity associated with antibody isotype switch and antibody-dependent cellular cytotoxicity activities. These results support possible clinical trials of the combination of DNA-EP/Ad-based cancer vaccines and IMO.