MilliporeSigma
  • Differences in the frequency of macrophage and T cell markers between focal and crescentic classes of anti-neutrophil cytoplasmic antibody (ANCA)-associated glomerulonephritis.
Differences in the frequency of macrophage and T cell markers between focal and crescentic classes of anti-neutrophil cytoplasmic antibody (ANCA)-associated glomerulonephritis.
Journal of nephropathology (2017-05-12)
Dana Kidder, Susan E Bray, Stewart Fleming
ABSTRACT
Anti-neutrophil cytoplasmic antibody (ANCA)-associated glomerulonephritis (AAGN) can be classified into; focal, crescentic, mixed and sclerotic classes. Macrophages and T lymphocytes are key players in mediating renal injury. The frequency of macrophage and T lymphocytes in different histological classes is unclear. We examined the frequency of macrophage and T lymphocyte markers in AAGN and assessed their correlation with renal function at presentation. Renal biopsies from 38 patients were included in immunohistochemistry analysis of macrophages (CD68, sialoadhesin [Sn] and mannose receptor [MR]) and T cells (CD4 and CD8) markers. The frequency of these markers in glomerular, periglomerular and interstitial compartments were measured in a blinded fashion. Biopsies were allocated a histological class of focal, crescentic, mixed or sclerotic. Scores were then matched to histological class and assessed for correlation with renal function. The biopsies were crescentic 19 (50%), focal 10 (26.3%), mixed 6 (15.7%) and sclerotic 3 (8%). Interstitial CD68+ macrophages and CD8+ T lymphocytes showed best correlation with renal function at the time of presentation. CD68+ macrophages were significantly increased in crescentic compared to focal AAGN. MR+ macrophages, CD4 and CD8 T cells were also elevated in the interstitium of crescentic compared to focal group. In this study interstitial CD68 and CD8 showed the highest association with the renal function at presentation. Differences in the cellular infiltrate between focal and crescentic AAGN were related to CD68+ macrophages and to interstitial MR+ macrophages and T lymphocytes. Further studies are needed to assess these differences across all four histological categories.