MilliporeSigma
  • ABCG2 is upregulated in Alzheimer's brain with cerebral amyloid angiopathy and may act as a gatekeeper at the blood-brain barrier for Abeta(1-40) peptides.
ABCG2 is upregulated in Alzheimer's brain with cerebral amyloid angiopathy and may act as a gatekeeper at the blood-brain barrier for Abeta(1-40) peptides.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2009-05-01)
Huaqi Xiong, Debbie Callaghan, Aimee Jones, Jianying Bai, Ingrid Rasquinha, Catherine Smith, Ke Pei, Douglas Walker, Lih-Fen Lue, Danica Stanimirovic, Wandong Zhang
ABSTRACT
Alzheimer's disease (AD) is characterized by accumulation and deposition of Abeta peptides in the brain. Abeta deposition in cerebrovessels occurs in many AD patients and results in cerebral amyloid angiopathy (AD/CAA). Since Abeta can be transported across blood-brain barrier (BBB), aberrant Abeta trafficking across BBB may contribute to Abeta accumulation in the brain and CAA development. Expression analyses of 273 BBB-related genes performed in this study showed that the drug transporter, ABCG2, was significantly upregulated in the brains of AD/CAA compared with age-matched controls. Increased ABCG2 expression was confirmed by Q-PCR, Western blot, and immunohistochemistry. Abcg2 was also increased in mouse AD models, Tg-SwDI and 3XTg. Abeta alone or in combination with hypoxia/ischemia failed to stimulate ABCG2 expression in BBB endothelial cells; however, conditioned media from Abeta-activated microglia strongly induced ABCG2 expression. ABCG2 protein in AD/CAA brains interacted and coimmunoprecipitated with Abeta. Overexpression of hABCG2 reduced drug uptake in cells; however, interaction of Abeta(1-40) with ABCG2 impaired ABCG2-mediated drug efflux. The role of Abcg2 in Abeta transport at the BBB was investigated in Abcg2-null and wild-type mice after intravenous injection of Cy5.5-labeled Abeta(1-40) or scrambled Abeta(40-1). Optical imaging analyses of live animals and their brains showed that Abcg2-null mice accumulated significantly more Abeta in their brains than wild-type mice. The finding was confirmed by immunohistochemistry. These results suggest that ABCG2 may act as a gatekeeper at the BBB to prevent blood Abeta from entering into brain. ABCG2 upregulation may serve as a biomarker of CAA vascular pathology in AD patients.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid