MilliporeSigma
  • Epitope recognition in the human-pig comparison model on fixed and embedded material.
Epitope recognition in the human-pig comparison model on fixed and embedded material.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (2015-07-26)
Carla Rossana Scalia, Rossella Gendusa, Maria Basciu, Lorella Riva, Lorenza Tusa, Antonella Musarò, Silvio Veronese, Angelo Formenti, Donatella D'Angelo, Angela Gabriella Ronzio, Giorgio Cattoretti, Maddalena Maria Bolognesi
ABSTRACT
The conditions and the specificity by which an antibody binds to its target protein in routinely fixed and embedded tissues are unknown. Direct methods, such as staining in a knock-out animal or in vitro peptide scanning of the epitope, are costly and impractical. We aimed to elucidate antibody specificity and binding conditions using tissue staining and public genomic and immunological databases by comparing human and pig-the farmed mammal evolutionarily closest to humans besides apes. We used a database of 146 anti-human antibodies and found that antibodies tolerate partially conserved amino acid substitutions but not changes in target accessibility, as defined by epitope prediction algorithms. Some epitopes are sensitive to fixation and embedding in a species-specific fashion. We also find that half of the antibodies stain porcine tissue epitopes that have 60% to 100% similarity to human tissue at the amino acid sequence level. The reason why the remaining antibodies fail to stain the tissues remains elusive. Because of its similarity with the human, pig tissue offers a convenient tissue for quality control in immunohistochemistry, within and across laboratories, and an interesting model to investigate antibody specificity.