MilliporeSigma
  • In vitro and in vivo anti-uveal melanoma activity of JSL-1, a novel HDAC inhibitor.
In vitro and in vivo anti-uveal melanoma activity of JSL-1, a novel HDAC inhibitor.
Cancer letters (2017-04-30)
Yun Wang, Maoxing Liu, Yanli Jin, Sheng Jiang, Jingxuan Pan
ABSTRACT
Uveal melanoma (UM) is the most common intraocular malignant neoplasm in adults. Despite the availability of enucleation, radiation and chemotherapy, the prognosis of patients with metastasis remains poor. Therefore, novel effective therapies for patients with metastatic UM are urgently needed. In the present study, we demonstrated that JSL-1, a novel HDAC inhibitor, effectively inhibited the proliferation. JSL-1 induced apoptosis with increased expression of proapoptotic BH3-only protein BIM in UM cells. JSL-1 suppressed migration and invasion of UM cells with MMP-2 decreased. Furthermore, JSL-1 blocked the canonical Wnt/β-catenin pathway, impaired self-renewal capacity and decreased percentage of ALDH