MilliporeSigma
Alpha
Cellular signalling (2017-03-18)
Lilian Fedra Castillo, Ezequiel M Rivero, Vincent Goffin, Isabel Alicia Lüthy
ABSTRACT
Breast cancer is the most frequent malignancy among women worldwide. We have described the expression of α