MilliporeSigma
  • Cavin-1 deficiency modifies myocardial and coronary function, stretch responses and ischaemic tolerance: roles of NOS over-activity.
Cavin-1 deficiency modifies myocardial and coronary function, stretch responses and ischaemic tolerance: roles of NOS over-activity.
Basic research in cardiology (2017-03-28)
Mika Kaakinen, Melissa E Reichelt, Zhibin Ma, Charles Ferguson, Nick Martel, Enzo R Porrello, James E Hudson, Walter G Thomas, Robert G Parton, John P Headrick
ABSTRACT
Caveolae and associated cavin and caveolins may govern myocardial function, together with responses to mechanical and ischaemic stresses. Abnormalities in these proteins are also implicated in different cardiovascular disorders. However, specific roles of the cavin-1 protein in cardiac and coronary responses to mechanical/metabolic perturbation remain unclear. We characterised cardiovascular impacts of cavin-1 deficiency, comparing myocardial and coronary phenotypes and responses to stretch and ischaemia-reperfusion in hearts from cavin-1
Sigma-Aldrich
Human TNNI3 / Troponin I, Cardiac Muscle ELISA Kit