MilliporeSigma
  • An improved glycerol biosensor with an Au-FeS-NAD-glycerol-dehydrogenase anode.
An improved glycerol biosensor with an Au-FeS-NAD-glycerol-dehydrogenase anode.
Biosensors & bioelectronics (2016-11-12)
Aishwarya Mahadevan, Sandun Fernando
ABSTRACT
An improved glycerol biosensor was developed via direct attachment of NAD
Sigma-Aldrich
Glycerol Dehydrogenase from Cellulomonas sp., lyophilized powder, ≥50 units/mg protein
Sigma-Aldrich
3-Aminophenylboronic acid monohydrate, 98%