MilliporeSigma
  • Induction of cell differentiation activates transcription of the Sarco/Endoplasmic Reticulum calcium-ATPase 3 gene (ATP2A3) in gastric and colon cancer cells.
Induction of cell differentiation activates transcription of the Sarco/Endoplasmic Reticulum calcium-ATPase 3 gene (ATP2A3) in gastric and colon cancer cells.
Molecular carcinogenesis (2016-07-20)
Lucía Flores-Peredo, Gabriela Rodríguez, Angel Zarain-Herzberg
ABSTRACT
The Sarco/Endoplasmic Reticulum Ca
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Iscove′s Modified Dulbecco′s Medium, With L-glutamine and 25 mM HEPES, without sodium bicarbonate, powder, suitable for cell culture