MilliporeSigma
  • Serotonin 5-HT2A and 5-HT6 receptors in the prefrontal cortex of Alzheimer and normal aging patients.
Serotonin 5-HT2A and 5-HT6 receptors in the prefrontal cortex of Alzheimer and normal aging patients.
BMC neuroscience (2006-04-28)
Dietrich E Lorke, Gang Lu, Eric Cho, David T Yew
ABSTRACT
It has been hypothesized that alterations of the serotonergic system contribute to neuropsychiatric symptoms in Alzheimer disease (AD). Cellular expressions of the two serotonergic receptors 5-HT2A and 5-HT6 have therefore been determined by immunohistochemistry in the prefrontal cortex of patients with AD (n=6) and normal age-matched controls (n = 7). In normal aging patients, 5-HT2A label was mainly observed in large pyramidal cells, but to a lesser extent also in small pyramidal cells and in stellate cells of cortical layers II-VI. In AD, a similar distribution was observed, but density of positive cells was significantly reduced by 33%. In aging control patients, the 5-HT6 receptor was expressed by pyramidal cells and occasional stellate cells, not only of layers II-V, but also of layer I, where a distinct label was observed in neurons and surrounding fibers. 5-HT6 receptor expression in AD patients had the same pattern, but was significantly decreased by 40%. Our results indicate that a decline in neurons expressing 5-HT2A, but also 5-HT6 receptors may play a role in the etiopathology of neuropsychiatric symptoms in AD.