MilliporeSigma
  • Humoral response in Toscana virus acute neurologic disease investigated by viral-protein-specific immunoassays.
Humoral response in Toscana virus acute neurologic disease investigated by viral-protein-specific immunoassays.
Clinical and diagnostic laboratory immunology (1999-01-05)
F Magurano, L Nicoletti
ABSTRACT
The Toscana virus (family Bunyaviridae, genus Phlebovirus) is the only sandfly-transmitted virus that demonstrates neurotropic activity. Clinical cases ranging from aseptic meningitis to meningoencephalitis caused by Toscana virus are yearly observed in central Italy during the summer, and several cases have been reported among tourists returning from zones of endemicity (Italy, Portugal, Spain, and Cyprus). In Toscana virus patients, immunoglobulin M (IgM) antibodies, usually present at the onset of symptoms, can reveal elevated titers by enzyme-linked immunosorbent assay and can persist for at least 1 year. IgG antibodies can be absent at the onset of symptoms: titers rise in convalescent sera and persist for many years. At least five proteins have been identified in Toscana virus-infected cells: nucleoprotein N, glycoproteins G1 and G2, a large protein (L) assumed to be a component of the polymerase, and two nonstructural proteins, NSm and NSs. We report results of a study on the antibody response to individual viral proteins in patients with Toscana virus-associated acute neurologic disease. Immunoblotting and semiquantitative radioimmunoprecipitation assay (RIPA) allow identification of nucleoprotein N as the major antigen responsible for both IgM and IgG responses. Antibodies to proteins other than nucleoprotein N are detected only by RIPA. Antibodies to glycoproteins are detected in about one-third of patients, and whereas their presence always predicts neutralization, some serum samples with neutralizing activity have undetectable levels of antibodies to G1-G2. Antibodies to nonstructural proteins NSm and NSs are also identified. The results obtained raise some questions about antigenic variability and relevant neutralization epitopes of Toscana virus.