MilliporeSigma
A possible role for metalloproteinases in renal cyst development.
American journal of physiology. Renal physiology (2001-02-22)
N Obermüller, N Morente, B Kränzlin, N Gretz, R Witzgall
ABSTRACT
The expansion of cysts in polycystic kidneys bears several similarities to the invasion of the extracellular matrix by benign tumors. We therefore hypothesized that cyst-lining epithelial cells produce extracellular matrix-degrading metalloproteinases and that the inhibition of these enzymes may represent a potential target for therapeutic intervention. Using in situ hybridization, we first analyzed the expression of membrane-type metalloproteinase 1 (MMP-14), an essential matrix metalloproteinase, of its inhibitor TIMP-2, and of the cytokine transforming growth factor (TGF)-beta2 in the (cy/+) rat model of autosomal-dominant polycystic kidney disease. Upregulated MMP-14 mRNA was predominantly located in cyst-lining epithelia and distal tubules, whereas TIMP-2 mRNA was confined almost exclusively to fibroblasts. TGF-beta2, a cytokine known to regulate the expression of matrix metalloproteinases and their inhibitors, was also expressed by cyst wall epithelia. We then treated (cy/+) rats with the metalloproteinase inhibitor batimastat for a period of 8 wk. The treatment with the metalloproteinase inhibitor batimastat resulted in a significant reduction of cyst number and kidney weight. Our study suggests that metalloproteinase inhibitors represent a new therapeutic tool against polycystic kidney disease, which should be applicable independently of the background of the disease.