MilliporeSigma
  • Isolation and characterization of histone H3 lysine 4 demethylase-containing complexes.
Isolation and characterization of histone H3 lysine 4 demethylase-containing complexes.
Methods (San Diego, Calif.) (2006-11-15)
Min Gyu Lee, Christopher Wynder, Jessica Norman, Ramin Shiekhattar
ABSTRACT
Histone methylation is involved in the regulation of many cellular processes. In the past 2 years, several histone demethylases including BHC110/LSD1 have been characterized. BHC110, the first known histone lysine demethylase, removes methyl groups from methylated histone H3 lysine 4 and has been found in many multi-protein complexes. Using one-step affinity purification, we have isolated enzymatically active BHC110-containing complexes. Here, we detail the methods used for the isolation and characterization of these histone demethylase complexes from a human stable cell line.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit