MilliporeSigma
  • Expression of glutamate decarboxylase isoform, GAD65, in human mononuclear leucocytes: a possible implication of C-terminal end deletion by Western blot and RT-PCR study.
Expression of glutamate decarboxylase isoform, GAD65, in human mononuclear leucocytes: a possible implication of C-terminal end deletion by Western blot and RT-PCR study.
Journal of biochemistry (2007-10-30)
Satoko Matsukawa, Hiroshi Ueno
ABSTRACT
Human peripheral blood leucocyte was examined for the expression of glutamate decarboxylase (GAD). Peripheral blood from healthy individuals was fractionated into polynuclear leucocytes and mononuclear leucocytes. Cell lysate from the mononuclear leucocytes was analysed by SDS-PAGE and Western blot. With antibody raised against unique C-terminal sequence for GAD65, two protein bands at 30 and 80 kDa were detected. However, with anti-GAD65/67 antibody recognizing very end of C-terminal, about 40 residues toward C-terminal, no protein bands were observed. Expression of mRNA coding for the epitope of these two antibodies was examined by using PCR technique. Results showed an evidence that mononuclear leucocytes express GAD65 with its C-terminal segment truncated. Our results have suggested an expression of GAD with the novel molecular weight may be caused by possible mononuclear leucocyte specific splicing errors.