MilliporeSigma
Targeting of the protein interaction site between FAK and IGF-1R.
Biochemical and biophysical research communications (2009-08-12)
Donghang Zheng, Elena Kurenova, Deniz Ucar, Vita Golubovskaya, Andrew Magis, David Ostrov, William G Cance, Steven N Hochwald
ABSTRACT
The interaction of focal adhesion kinase (FAK) and insulin-like growth factor-1 receptor (IGF-1R) plays an important role in cancer cell survival. Targeting this interaction with small molecule drugs could be a novel strategy in cancer therapy. By a series of pull-down assays using GST-tagged FAK fragments and His-tagged IGF-1R intracellular fragments, we showed that the FAK-NT2 (a.a. 127-243) domain directly interacts with the N-terminal part of the IGF-1R intracellular domain. Overexpressed FAK-NT2 domain was also shown to co-localize with IGF-1R in pancreatic cells. Computational modeling was used to predict the binding configuration of these two domains and to screen for small molecules binding to the interaction site. This strategy successfully identified a lead compound that disrupts FAK/IGF-1R interaction.
Sigma-Aldrich
Monoclonal Anti-Glutathione-S-Transferase (GST) antibody produced in mouse, clone GST-2, ascites fluid
Sigma-Aldrich
Anti-IGF-IR (Ab-1) Mouse mAb (αIR3), liquid, clone αIR3, Calbiochem®