MilliporeSigma
  • Increased water intake decreases progression of polycystic kidney disease in the PCK rat.
Increased water intake decreases progression of polycystic kidney disease in the PCK rat.
Journal of the American Society of Nephrology : JASN (2006-06-30)
Shizuko Nagao, Kazuhiro Nishii, Makoto Katsuyama, Hiroki Kurahashi, Tohru Marunouchi, Hisahide Takahashi, Darren P Wallace
ABSTRACT
Renal enlargement in polycystic kidney disease (PKD) is caused by the proliferation of mural epithelial cells and transepithelial fluid secretion into the cavities of innumerable cysts. Arginine vasopressin (AVP) stimulates the proliferation of human PKD cells in vitro via cAMP-dependent activation of the B-Raf/MEK (MAPK/ERK kinase/extracellular signal-regulated kinase (ERK) pathway. ERK activity is elevated in cells that line the cysts in animals with PKD, and AVP receptor antagonists reduce ERK activity and halt disease progression. For suppression of the effect of AVP physiologically, water intake was increased in PCK rats, a model of PKD, and the effect on renal morphology, cellular mechanism, and function was determined. The addition of 5% glucose in the drinking water increased fluid intake approximately 3.5-fold compared with rats that received tap water. In PCK rats, increased water intake for 10 wk reduced urinary AVP excretion (68.3%), and urine osmolality fell below 290 mOsmol/kg. High water intake was associated with reduced renal expression of AVP V2 receptors (41.0%), B-Raf (15.4%), phosphorylated ERK (38.1%), and proliferating cell nuclear antigen-positive renal cells (61.7%). High water intake reduced the kidney/body weight ratio 28.0% and improved renal function. Taken together, these data demonstrate that water intake that is sufficient to cause persistent water diuresis suppresses B-Raf/MEK/ERK activity and decreases cyst and renal volumes in PCK rats. It is suggested that limiting serum AVP levels by increased water intake may be beneficial to some patients with PKD.
Sigma-Aldrich
Anti-MAP Kinase (ERK-1, ERK-2) antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Monoclonal Anti-Proliferating Cell Nuclear Antigen antibody produced in mouse, clone PC 10, ascites fluid