MilliporeSigma
  • Plasminogen activator inhibitor-1 aids nerve growth factor-induced differentiation and survival of pheochromocytoma cells by activating both the extracellular signal-regulated kinase and c-Jun pathways.
Plasminogen activator inhibitor-1 aids nerve growth factor-induced differentiation and survival of pheochromocytoma cells by activating both the extracellular signal-regulated kinase and c-Jun pathways.
Neuroscience (2006-05-09)
S Soeda, K Shinomiya, T Ochiai, S Koyanagi, A Toda, R Eyanagi, H Shimeno
ABSTRACT
Astrocytes are thought to be critical to neurons' surviving damage caused by ischemic stroke or other injury. Plasminogen activator inhibitor-1 is one of the active soluble factors released by astrocytes and regulates plasminogen activator-plasmin proteolytic sequence in the CNS as a serpin. In this study, we show that plasminogen activator inhibitor-1 can promote neurite outgrowth and survival of rat pheochromocytoma cells in serum-deprived conditions, and that this neuroprotective activity is correlated with enhanced activation of both extracellular signal-regulated kinases following a direct phosphorylation of nerve growth factor receptor, Trk A, and of c-Jun. Our results suggest that plasminogen activator inhibitor-1 can act as a neurotrophic factor, protecting neurons from serum deprivation-induced neuron death not only by compensating for nerve growth factor functions, but also by activating the c-Jun/activating protein-1 pathway.