MilliporeSigma
  • Block in development at the pre-B-II to immature B cell stage in mice without Ig kappa and Ig lambda light chain.
Block in development at the pre-B-II to immature B cell stage in mice without Ig kappa and Ig lambda light chain.
Journal of immunology (Baltimore, Md. : 1950) (2003-01-23)
Xiangang Zou, Tony A Piper, Jennifer A Smith, Nicholas D Allen, Jian Xian, Marianne Brüggemann
ABSTRACT
Silencing individual C (constant region) lambda genes in a kappa(-/-) background reduces mature B cell levels, and L chain-deficient (lambda(-/-)kappa(-/-)) mice attain a complete block in B cell development at the stage when L chain rearrangement, resulting in surface IgM expression, should be completed. L chain deficiency prevents B cell receptor association, and L chain function cannot be substituted (e.g., by surrogate L chain). Nevertheless, precursor cell levels, controlled by developmental progression and checkpoint apoptosis, are maintained, and B cell development in the bone marrow is fully retained up to the immature stage. L chain deficiency allows H chain retention in the cytoplasm, but prevents H chain release from the cell, and as a result secondary lymphoid organs are B cell depleted while T cell levels remain normal.