MilliporeSigma
  • Apamin, a highly potent fetal L-type Ca2+ current blocker in single heart cells.
Apamin, a highly potent fetal L-type Ca2+ current blocker in single heart cells.
The American journal of physiology (1992-02-01)
G Bkaily, A Sculptoreanu, D Jacques, D Economos, D Ménard
ABSTRACT
Apamin, a bee venom polypeptide, was reported to block the naturally occurring Ca2+ slow action potentials (APs) in cultured cell reaggregates from old chick hearts [Bkaily, G. et al. Am. J. Physiol. 248 (Heart Circ. Physiol. 17): H961-H965, 1985] as well as the tetrodotoxin (TTX)- and Mn(2+)-insensitive slow Na+ current in young embryonic chick heart cells (Bkaily, G. In Vitro Toxicology. Academic, In press; Bkaily et al. J. Mol. Cell. Cardiol. 23: 25-39, 1991). With the use of the whole cell voltage-clamp technique in single ventricular cells from 10-day-old chick embryos and 17- to 20-wk-old human fetuses, two types of Ca2+ currents (ICa), T and L, were found. These two types of slow inward current in both heart preparations were nearly similar in their voltage, kinetics, and pharmacology. Apamin, a slow Ca2+ action potential blocker in old embryonic chick heart, was found to block the L-type ICa (IL) in a dose-dependent manner without affecting the T-type ICa in both heart cell preparations. The blockade of the IL by apamin was completely reversible upon washout with apamin-free solution. Therefore, when compared with nifedipine or to PN 200-110, apamin seems to be a highly potent L-type Ca2+ channel blocker in heart cells.