MilliporeSigma
  • CGA-7 and HHF, two monoclonal antibodies that recognize muscle actin and react with adherent cells in human long-term bone marrow cultures.
CGA-7 and HHF, two monoclonal antibodies that recognize muscle actin and react with adherent cells in human long-term bone marrow cultures.
Blood (1985-11-01)
P Charbord, A M Gown, A Keating, J W Singer
ABSTRACT
The CGA-7, a monoclonal antibody that reacts with smooth muscle cell actin but not with endothelial cell or fibroblast actin, and HHF, a monoclonal antibody that reacts with smooth muscle, skeletal muscle, and cardiac muscle actin, both recognize microfilaments present within adherent cells from actively hematopoietic human long-term marrow cultures. Macrophages, monocytes, and cultured marrow fibroblasts do not react with either antibody. These data suggest that the anti-actin antibodies may serve as useful markers for in vitro microenvironmental cells and lend support to the hypothesis that stromal cells from long-term marrow cultures are different from marrow fibroblasts and may constitute a unique cell lineage.