MilliporeSigma
  • Dissection of transcriptional and cis-regulatory control of differentiation in human pancreatic cancer.
Dissection of transcriptional and cis-regulatory control of differentiation in human pancreatic cancer.
The EMBO journal (2016-01-16)
Giuseppe R Diaferia, Chiara Balestrieri, Elena Prosperini, Paola Nicoli, Paola Spaggiari, Alessandro Zerbi, Gioacchino Natoli
ABSTRACT
The histological grade of carcinomas describes the ability of tumor cells to organize in differentiated epithelial structures and has prognostic and therapeutic impact. Here, we show that differential usage of the genomic repertoire of transcriptional enhancers leads to grade-specific gene expression programs in human pancreatic ductal adenocarcinoma (PDAC). By integrating gene expression profiling, epigenomic footprinting, and loss-of-function experiments in PDAC cell lines of different grade, we identified the repertoires of enhancers specific to high- and low-grade PDACs and the cognate set of transcription factors acting to maintain their activity. Among the candidate regulators of PDAC differentiation, KLF5 was selectively expressed in pre-neoplastic lesions and low-grade primary PDACs and cell lines, where it maintained the acetylation of grade-specific enhancers, the expression of epithelial genes such as keratins and mucins, and the ability to organize glandular epithelia in xenografts. The identification of the transcription factors controlling differentiation in PDACs will help clarify the molecular bases of its heterogeneity and progression.
Sigma-Aldrich
Anti-ELF3 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-FOS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-HNF1B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution