MilliporeSigma
  • International Union of Pharmacology. XXXIX. Compendium of voltage-gated ion channels: sodium channels.
International Union of Pharmacology. XXXIX. Compendium of voltage-gated ion channels: sodium channels.
Pharmacological reviews (2003-12-06)
William A Catterall, Alan L Goldin, Stephen G Waxman
ABSTRACT
This summary article presents an overview of the molecular relationships among the voltage-gated sodium channels and a standard nomenclature for them, which is derived from the IUPHAR Compendium of Voltage-Gated Ion Channels. The complete Compendium, including data tables for each member of the sodium channel family can be found at <http://www.iuphar-db.org/iuphar-ic/>.