MilliporeSigma
  • Human puromycin-sensitive aminopeptidase: cloning of 3' UTR, evidence for a polymorphism at a.a. 140 and refined chromosomal localization to 17q21.
Human puromycin-sensitive aminopeptidase: cloning of 3' UTR, evidence for a polymorphism at a.a. 140 and refined chromosomal localization to 17q21.
Cytogenetics and cell genetics (2001-07-04)
W O Bauer, I Nanda, G Beck, M Schmid, F Jakob
ABSTRACT
Puromycin-sensitive aminopeptidase is a predominantly cytoplasmatic zinc-dependent exopeptidase. Its physiological function is not known to date. Here we report data on tissue distribution, a polymorphism within the coding region and the complete 3' UTR. The gene (NPEPPS alias PSA) was physically mapped to chromosome 17q21.2-->q21.32 using fluorescence in situ hybridization.