MilliporeSigma
  • A Genome-Wide Association Study Identifies the Skin Color Genes IRF4, MC1R, ASIP, and BNC2 Influencing Facial Pigmented Spots.
A Genome-Wide Association Study Identifies the Skin Color Genes IRF4, MC1R, ASIP, and BNC2 Influencing Facial Pigmented Spots.
The Journal of investigative dermatology (2015-02-24)
Leonie C Jacobs, Merel A Hamer, David A Gunn, Joris Deelen, Jaspal S Lall, Diana van Heemst, Hae-Won Uh, Albert Hofman, André G Uitterlinden, Christopher E M Griffiths, Marian Beekman, P Eline Slagboom, Manfred Kayser, Fan Liu, Tamar Nijsten
ABSTRACT
Facial pigmented spots are a common skin aging feature, but genetic predisposition has yet to be thoroughly investigated. We conducted a genome-wide association study for pigmented spots in 2,844 Dutch Europeans from the Rotterdam Study (mean age: 66.9±8.0 years; 47% male). Using semi-automated image analysis of high-resolution digital facial photographs, facial pigmented spots were quantified as the percentage of affected skin area (mean women: 2.0% ±0.9, men: 0.9% ±0.6). We identified genome-wide significant association with pigmented spots at three genetic loci: IRF4 (rs12203592, P=1.8 × 10(-27)), MC1R (compound heterozygosity score, P=2.3 × 10(-24)), and RALY/ASIP (rs6059655, P=1.9 × 10(-9)). In addition, after adjustment for the other three top-associated loci the BNC2 locus demonstrated significant association (rs62543565, P=2.3 × 10(-8)). The association signals observed at all four loci were successfully replicated (P<0.05) in an independent Dutch cohort (Leiden Longevity Study n=599). Although the four genes have previously been associated with skin color variation and skin cancer risk, all association signals remained highly significant (P<2 × 10(-8)) when conditioning the association analyses on skin color. We conclude that genetic variations in IRF4, MC1R, RALY/ASIP, and BNC2 contribute to the acquired amount of facial pigmented spots during aging, through pathways independent of the basal melanin production.