MilliporeSigma
  • Peripheral blood hematopoietic stem and progenitor cell frequency is unchanged in patients with alpha-1-antitrypsin deficiency.
Peripheral blood hematopoietic stem and progenitor cell frequency is unchanged in patients with alpha-1-antitrypsin deficiency.
International journal of hematology (2014-04-23)
Evert-Jan F M de Kruijf, Gonnie M Alkemade, Ronald van Os, Willem E Fibbe, Melissa van Pel
ABSTRACT
Granulocyte-colony-stimulating factor (G-CSF)-induced hematopoietic stem and progenitor cell (HSPC) mobilization is associated with the release of neutrophil-derived proteases. Previously, we have shown that alpha-1-antitrypsin (AAT) inhibits these proteases in mice, resulting in inhibition of HSPC mobilization. Here, we studied the relationship between AAT and HSPC in steady state and cytokine-induced mobilization in humans. Patients with alpha-1-antitrypsin deficiency (AATD) have an 85-90 % decrease of AAT in the peripheral blood (PB). We hypothesized that this leads to increased proteolytic activity in the bone marrow and increased steady-state PB HSPC numbers. Using flow cytometry and semi-solid cell culture, we found no significant difference in PB HSPC in AATD patients (n = 18) as compared to controls (n = 22). Healthy stem cell donors (n = 43) were mobilized with G-CSF for 5 days and the number of CD45(+)/CD34(+) HSPC were determined in PB. We found that, during mobilization, PB AAT levels increased significantly, positively correlating with PB CD45(+)/CD34(+) cells (r = 0.31, p = 0.005). In conclusion, although serum AAT levels and HSPC mobilization in healthy stem cell donors are positively correlated, AAT is not an indispensable protease-inhibitor in the constitutive circulation of HSPC. These findings suggest a model in which both protease-dependent and -independent pathways contribute to HSPC mobilization.
Sigma-Aldrich
G-CSF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
Granulocyte colony-stimulating factor human, G-CSF, recombinant, expressed in E. coli, suitable for cell culture