MilliporeSigma
  • Inhibition of inducible nitric oxide synthase prevents shock wave therapy induced renal injury.
Inhibition of inducible nitric oxide synthase prevents shock wave therapy induced renal injury.
Renal failure (2014-02-12)
Bilal Firat Alp, Ercan Malkoc, Zafer Demirer, Ali Guragac, Turker Turker, Ertan Altayli, Ayhan Ozcan, Bulent Uysal, Turgut Topal, Emin Ozgur Akgul, Ibrahim Yildirim, Ahmet Guven
ABSTRACT
Shock wave lithotripsy treatment (SWT) is not completely free from side effects; one of the accused mechanisms for renal injury during SWT is oxygen- and nitrogen-derived free radical productions. Therefore, we aimed to evaluate the effect of inhibition of nitric oxide (NO) production by N-[3(aminomethyl) benzyl) acetamidine] (1400W), highly selective inducible nitric oxide synthase (iNOS) inhibitor, at SWT-induced kidney damage. Twenty-four rats those underwent right nephrectomy procedure were divided equally into three groups as control, SWT, and SWT + 1400W. 1400W was administered at a dose of 10 mg/kg at 2 h prior to SWT procedure and at the beginning of SWT procedure via intraperitoneal route and continued daily for consecutive 3 days. At the end of the fourth day, animals were killed via decapitation and trunk blood and the left kidneys were taken for biochemical and histopathologic evaluation. SWT caused renal tubular damage and increased lipid peroxidation and antioxidant enzyme activities and SWT also significantly increased nitro-oxidative products. Inhibition of iNOS via administration of 1400W ameliorated renal injury and decreased tissue lipid peroxidation (malondialdehyde), superoxide dismutase, glutathione peroxidase and nitrite/nitrate levels (NOx). In addition, it was seen that histolopathological changes were attenuated in the SWT + 1400W group when compared to SWT group. SWT-induced renal injury might be due to excessive production of oxygen free radicals and NO production. Inhibition of iNOS attenuates renal injury following SWT treatment. It can be concluded that iNOS inhibitors or peroxynitrite scavengers might be used to protect the kidneys against SWT-induced morphological and functional injuries.