MilliporeSigma
  • Matrix metalloproteinase-14 expression and its prognostic value in cervical carcinoma.
Matrix metalloproteinase-14 expression and its prognostic value in cervical carcinoma.
Cell biochemistry and biophysics (2014-05-03)
Huayi Wang, Xianhua Zhang, Liming Huang, Jia Li, Shuyun Qu, Fenglian Pan
ABSTRACT
The objective of the study was to evaluate the expression of matrix metalloproteinase-14 (MMP-14) in cervical carcinoma and correlate its expression with clinicopathological parameters, recurrence, and survival of the patients. The expressions of MMP-14 in normal cervical mucosa and cervical carcinoma tissue were detected with immunohistochemistry. Survival analysis by the Kaplan-Meier method was performed to assess prognostic significance. The positive expression rate of MMP-14 in cervical carcinoma tissue was 81.6 %(111/136), and there was significant difference on their positive expression rates between in cervical carcinoma tissue and in normal cervical mucosa(22.4 %)(13/58)(P < 0.05);The positive expression rates of MMP-14 in patients with poor histologic differentiation, lymph node metastasis, and recurrence group were heightened. Using Kaplan-Meier analysis, a comparison of survival curves of low versus high expressions of MMP-14 revealed a highly significant difference in human cervical carcinoma tissue (P < 0.05), which suggests that overexpression of MMP-14 is associated with a worse prognosis. The MMP-14 which promotes angiogenes is associated with lymph node metastasis, vascular invasion, and poor prognosis of cervical carcinoma. The current study shows that MMP-14 may be an independent prognostic factor for cervical carcinoma patients.