MilliporeSigma
  • Brief Report: Involvement of TNFRSF11A molecular defects in autoinflammatory disorders.
Brief Report: Involvement of TNFRSF11A molecular defects in autoinflammatory disorders.
Arthritis & rheumatology (Hoboken, N.J.) (2014-06-04)
Isabelle Jéru, Emmanuelle Cochet, Philippe Duquesnoy, Véronique Hentgen, Bruno Copin, Maria Teresa Mitjavila-Garcia, Shayan Sheykholeslami, Gaëlle Le Borgne, Florence Dastot-Le Moal, Valérie Malan, Sonia Karabina, Mathieu Mahevas, Sandra Chantot-Bastaraud, Jean-Claude Lecron, Laurence Faivre, Serge Amselem
ABSTRACT
Autoinflammatory disorders are caused by a primary dysfunction of the innate immune system. Among these disorders are hereditary recurrent fevers, which are characterized by recurrent episodes of fever and inflammatory manifestations affecting multiple tissues. Hereditary recurrent fevers often lack objective diagnostic criteria, thereby hampering the identification of disease-causing genes. This study was undertaken to identify a gene responsible for hereditary recurrent fevers. Copy number variations and point mutations were sought by array-comparative genomic hybridization and polymerase chain reaction sequencing, respectively. Serum cytokine levels were measured using Luminex technology. The effect of TNFRSF11A molecular defects on NF-κB signaling in cells expressing wild-type and mutated forms of the receptor was evaluated by luciferase assay. A patient with multiple congenital anomalies and hereditary recurrent fever was found to carry a de novo heterozygous complex chromosomal rearrangement encompassing a duplication of TNFRSF11A, a gene known to regulate fever in rodents. We also identified a heterozygous frameshift mutation (p.Met416Cysfs*110) in TNFRSF11A in a mother and daughter with isolated hereditary recurrent fever. This mutation was associated with increased secretion of several inflammatory cytokines (tumor necrosis factor α [TNFα], interleukin-18 [IL-18], IL-1 receptor antagonist, interferon-γ) and altered the biologic effects of the receptor on NF-κB signaling. The disease in the patients described herein exhibits striking clinical similarities to TNF receptor-associated periodic syndrome, another hereditary recurrent fever involving a gene of the same family (TNFRSF1A). The involvement of TNFRSF11A in hereditary recurrent fever highlights the key role of this receptor in innate immunity. The present results also suggest that TNFRSF11A screening could serve as a new diagnostic test for autoinflammatory disorders.