MilliporeSigma
  • Pharmacokinetic and pharmacodynamic interaction between gemigliptin and metformin in healthy subjects.
Pharmacokinetic and pharmacodynamic interaction between gemigliptin and metformin in healthy subjects.
Clinical drug investigation (2014-03-15)
Dongseong Shin, Young Min Cho, SeungHwan Lee, Kyoung Soo Lim, Jeong-Ae Kim, Ji-Yung Ahn, Joo-Youn Cho, Howard Lee, In-Jin Jang, Kyung-Sang Yu
ABSTRACT
Gemigliptin is a novel dipeptidyl peptidase-4 (DPP-4) inhibitor used in the treatment of type 2 diabetes mellitus. This study evaluated possible pharmacodynamic and pharmacokinetic interactions between gemigliptin and metformin and investigated their tolerability. A randomized, open-label, multiple-dose, three-treatment, three-period, three-sequence crossover study was conducted in healthy male subjects. Twenty-seven subjects received gemigliptin (50 mg once daily), metformin (1,000 mg twice a day), or both drugs for 7 days per dosing period. Blood samples were drawn over 24 h on the seventh day of each period for pharmacokinetic and pharmacodynamic evaluations, including plasma DPP-4 activity and total/active glucagon-like peptide-1 (GLP-1) levels. Meal tolerance tests were conducted for pharmacodynamic assessment on the eighth day. Safety and tolerability were evaluated using adverse events, vital signs, ECGs, and clinical laboratory tests. Coadministration of gemigliptin and metformin had no significant effect on the pharmacokinetics of gemigliptin or metformin. The inhibition of DPP-4 by gemigliptin was not affected by coadministration with metformin. Co-therapy of gemigliptin and metformin showed additional effects by increasing plasma active GLP-1 concentrations and lowering serum glucose levels. The plasma glucagon level was lower in co-therapy than with metformin monotherapy. The coadministration of gemigliptin and metformin was well-tolerated without serious adverse events. Coadministration of gemigliptin and metformin showed beneficial anti-diabetic effects without pharmacokinetic drug-drug interactions.
Sigma-Aldrich
Mouse Granzyme B ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human GCG / Glucagon ELISA Kit