MilliporeSigma
  • Genetic analysis of an orbital metastasis from a primary hepatic neuroendocrine carcinoma.
Genetic analysis of an orbital metastasis from a primary hepatic neuroendocrine carcinoma.
Oncology reports (2014-07-23)
Jacob Ø Rasmussen, Sarah L von Holstein, Jan U Prause, Ben Vainer, Alastair B Hansen, André Fehr, Göran Stenman, Steffen Heegaard
ABSTRACT
A 71-year-old female with a known history of primary hepatic neuroendocrine carcinoma, presented with a visual defect, proptosis and restricted eye movements of the right eye. Biopsies from the orbit and from the primary hepatic neuroendocrine carcinoma showed similar morphological and immunohistochemical features, and high-resolution, array-based comparative genomic hybridization demonstrated loss of one copy each of chromosomes 3 and 18, and gain of 1q both in the primary hepatic neuroendocrine carcinoma and in the orbital tumour. The orbital mass was diagnosed as a metastasis from the primary hepatic neuroendocrine carcinoma. Primary hepatic neuroendocrine tumours are extremely rare, and the orbit is an extremely rare location for a neuroendocrine carcinoma metastasis. This is the first reported case of an orbital metastasis with origin from a primary hepatic neuroendocrine carcinoma.