MilliporeSigma
  • Efficient UV-induced charge separation and recombination in an 8-oxoguanine-containing dinucleotide.
Efficient UV-induced charge separation and recombination in an 8-oxoguanine-containing dinucleotide.
Proceedings of the National Academy of Sciences of the United States of America (2014-07-30)
Yuyuan Zhang, Jordan Dood, Ashley A Beckstead, Xi-Bo Li, Khiem V Nguyen, Cynthia J Burrows, Roberto Improta, Bern Kohler
ABSTRACT
During the early evolution of life, 8-oxo-7,8-dihydro-2'-deoxyguanosine (O) may have functioned as a proto-flavin capable of repairing cyclobutane pyrimidine dimers in DNA or RNA by photoinduced electron transfer using longer wavelength UVB radiation. To investigate the ability of O to act as an excited-state electron donor, a dinucleotide mimic of the FADH2 cofactor containing O at the 5'-end and 2'-deoxyadenosine at the 3'-end was studied by femtosecond transient absorption spectroscopy in aqueous solution. Following excitation with a UV pulse, a broadband mid-IR pulse probed vibrational modes of ground-state and electronically excited molecules in the double-bond stretching region. Global analysis of time- and frequency-resolved transient absorption data coupled with ab initio quantum mechanical calculations reveal vibrational marker bands of nucleobase radical ions formed by electron transfer from O to 2'-deoxyadenosine. The quantum yield of charge separation is 0.4 at 265 nm, but decreases to 0.1 at 295 nm. Charge recombination occurs in 60 ps before the O radical cation can lose a deuteron to water. Kinetic and thermodynamic considerations strongly suggest that all nucleobases can undergo ultrafast charge separation when π-stacked in DNA or RNA. Interbase charge transfer is proposed to be a major decay pathway for UV excited states of nucleic acids of great importance for photostability as well as photoredox activity.
Sigma-Aldrich
Adenosine 3′-monophosphate, from yeast