MilliporeSigma
  • 17-Dimethylaminoethylamino-17-demethoxygeldanamycin attenuates inflammatory responses in experimental stroke.
17-Dimethylaminoethylamino-17-demethoxygeldanamycin attenuates inflammatory responses in experimental stroke.
Biological & pharmaceutical bulletin (2014-11-05)
Jia Qi, Xiao Han, Hai Tao Liu, Ting Chen, Jin Lian Zhang, Ping Yang, Shu Hong Bo, Xiao Tong Lu, Jian Zhang
ABSTRACT
Heat shock protein 90 (HSP90) is a ubiquitous molecular chaperone involved in the proper conformation of many proteins. HSP90 inhibitors (17-dimethyl aminoethylamino-17-demethoxygeldanamycin hydrochloride [17-DMAG]) bind to and inactivate HSP90, suppressing some key signaling pathways involved in the inflammatory process. Since considerable evidence suggests that inflammation accounts for the progression of cerebral ischemic injury, we investigated whether 17-DMAG can modulate inflammatory responses in middle cerebral artery occluded (MCAO) mice. Male C57/BL6 mice were pretreated with 17-DMAG or vehicle for 7 d before being subjected to transient occlusion of middle cerebral artery and reperfusion. Mice were evaluated at 24 h after MCAO for neurological deficit scoring. Moreover, the mechanism of the anti-inflammatory effect of 17-DMAG was investigated with a focus on nuclear factor kappa B (NF-κB) pathway. 17-DMAG significantly reduced cerebral infarction and improved neurological outcome. 17-DMAG suppressed activation of microglia and decreased phosphorylation of inhibitory (I)κB and subsequent nuclear translocation of p65, which eventually downregulated expression of NF-κB-regulated genes. These results suggest that 17-DMAG has a promising therapeutic effect in ischemic stroke treatment through an anti-inflammatory mechanism.