MilliporeSigma
  • High levels of soluble CTLA-4 are present in anti-mitochondrial antibody positive, but not in antibody negative patients with primary biliary cirrhosis.
High levels of soluble CTLA-4 are present in anti-mitochondrial antibody positive, but not in antibody negative patients with primary biliary cirrhosis.
PloS one (2014-11-11)
Daniele Saverino, Giampaola Pesce, Princey Antola, Brunetta Porcelli, Ignazio Brusca, Danilo Villalta, Marilina Tampoia, Renato Tozzoli, Elio Tonutti, Maria Grazia Alessio, Marcello Bagnasco, Nicola Bizzaro
ABSTRACT
Primary biliary cirrhosis (PBC) is a chronic autoimmune cholestatic liver disease frequently characterized by anti-mitochondrial autoantibodies (AMA). A minority of patients are AMA-negative. Cytotoxic-T-Lymphocyte-Antigen-4 (CTLA-4) is a surface molecule expressed on activated T-cells delivering a critical negative immunoregulatory signal. A soluble form of CTLA-4 (sCTLA-4) has been detected at high concentrations in several autoimmune diseases, and its possible functional meaning has been suggested. We aimed to evaluate sCTLA-4 concentration in sera of patients with PBC and to correlate it to immunological abnormalities associated with the disease. Blood samples were collected from 82 PBC-patients diagnosed according to international criteria (44 AMA-positive/MIT3-positive and 38 AMA-negative-MIT3-negative), and 65 controls. sCTLA-4 levels were evaluated by ELISA and Western blot. Increased sCTLA-4 concentrations were found in all AMA-positive PBC-patients, but in none of the AMA-negative ones, nor in normal controls or in controls with unrelated liver diseases. sCTLA-4 presence was associated with autoantibodies against MIT3, but not with nuclear autoantibodies (sp100, gp210). This is the first study to demonstrate that levels of sCTLA-4 are elevated in sera of PBC patients. However, they are clearly restricted to patients with AMA positivity, suggesting an immunological difference with respect to AMA-negative ones.