MilliporeSigma
  • Ribosome profiling reveals a cell-type-specific translational landscape in brain tumors.
Ribosome profiling reveals a cell-type-specific translational landscape in brain tumors.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-08-15)
Christian Gonzalez, Jennifer S Sims, Nicholas Hornstein, Angeliki Mela, Franklin Garcia, Liang Lei, David A Gass, Benjamin Amendolara, Jeffrey N Bruce, Peter Canoll, Peter A Sims
ABSTRACT
Glioma growth is driven by signaling that ultimately regulates protein synthesis. Gliomas are also complex at the cellular level and involve multiple cell types, including transformed and reactive cells in the brain tumor microenvironment. The distinct functions of the various cell types likely lead to different requirements and regulatory paradigms for protein synthesis. Proneural gliomas can arise from transformation of glial progenitors that are driven to proliferate via mitogenic signaling that affects translation. To investigate translational regulation in this system, we developed a RiboTag glioma mouse model that enables cell-type-specific, genome-wide ribosome profiling of tumor tissue. Infecting glial progenitors with Cre-recombinant retrovirus simultaneously activates expression of tagged ribosomes and delivers a tumor-initiating mutation. Remarkably, we find that although genes specific to transformed cells are highly translated, their translation efficiencies are low compared with normal brain. Ribosome positioning reveals sequence-dependent regulation of ribosomal activity in 5'-leaders upstream of annotated start codons, leading to differential translation in glioma compared with normal brain. Additionally, although transformed cells express a proneural signature, untransformed tumor-associated cells, including reactive astrocytes and microglia, express a mesenchymal signature. Finally, we observe the same phenomena in human disease by combining ribosome profiling of human proneural tumor and non-neoplastic brain tissue with computational deconvolution to assess cell-type-specific translational regulation.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture