MilliporeSigma
  • Phenotypic overlap between familial exudative vitreoretinopathy and microcephaly, lymphedema, and chorioretinal dysplasia caused by KIF11 mutations.
Phenotypic overlap between familial exudative vitreoretinopathy and microcephaly, lymphedema, and chorioretinal dysplasia caused by KIF11 mutations.
JAMA ophthalmology (2014-08-16)
Johane M Robitaille, Roxanne M Gillett, Marissa A LeBlanc, Daniel Gaston, Mathew Nightingale, Michael P Mackley, Sandhya Parkash, Julie Hathaway, Aidan Thomas, Anna Ells, Elias I Traboulsi, Elise Héon, Mélanie Roy, Stavit Shalev, Conrad V Fernandez, Christine MacGillivray, Karin Wallace, Somayyeh Fahiminiya, Jacek Majewski, Christopher R McMaster, Karen Bedard
ABSTRACT
Retinal detachment with avascularity of the peripheral retina, typically associated with familial exudative vitreoretinopathy (FEVR), can result from mutations in KIF11, a gene recently identified to cause microcephaly, lymphedema, and chorioretinal dysplasia (MLCRD) as well as chorioretinal dysplasia, microcephaly, and mental retardation (CDMMR). Ophthalmologists should be aware of the range of presentations for mutations in KIF11 because the phenotypic distinction between FEVR and MLCRD/CDMMR portends management implications in patients with these conditions. To identify gene mutations in patients who present with a FEVR phenotype and explore the spectrum of ocular and systemic abnormalities caused by KIF11 mutations in a cohort of patients with FEVR or microcephaly in conjunction with chorioretinopathy or FEVR. Clinical data and DNA were collected from each participant between 1998 and 2013 from the clinical practices of ophthalmologists and clinical geneticists internationally. Twenty-eight FEVR probands with diagnoses made by the referring physician and without a known FEVR gene mutation, and 3 with microcephaly and chorioretinopathy, were included. At least 1 patient in each pedigree manifested 1 or more of the following: macular dragging, partial retinal detachment, falciform folds, or total retinal detachment. Whole-exome sequencing was conducted on affected members in multiplex pedigrees, and Sanger sequencing of the 22 exons of the KIF11 gene was performed on singletons. Clinical data and history were collected and reviewed. Identification of mutations in KIF11. Four novel heterozygous KIF11 mutations and 1 previously published mutation were identified in probands with FEVR: p.A218Gfs*15, p.E470X, p.R221G, c.790-1G>T, and the previously described heterozygous p.R47X. Documentation of peripheral avascular areas on intravenous fluorescein angiography was possible in 2 probands with fibrovascular proliferation demonstrating phenotypic overlap with FEVR. Mutations in KIF11 cause a broader spectrum of ocular disease than previously reported, including retinal detachment. The KIF11 gene likely plays a role in retinal vascular development and mutations in this gene can lead to clinical overlap with FEVR. Cases of FEVR should be carefully inspected for the presence of microcephaly as a marker for KIF11-related disease to enhance the accuracy of the prognosis and genetic counseling.