MilliporeSigma
  • Control of O-glycan branch formation. Molecular cloning of human cDNA encoding a novel beta1,6-N-acetylglucosaminyltransferase forming core 2 and core 4.
Control of O-glycan branch formation. Molecular cloning of human cDNA encoding a novel beta1,6-N-acetylglucosaminyltransferase forming core 2 and core 4.
The Journal of biological chemistry (1999-02-13)
T Schwientek, M Nomoto, S B Levery, G Merkx, A G van Kessel, E P Bennett, M A Hollingsworth, H Clausen
ABSTRACT
A novel human UDP-GlcNAc:Gal/GlcNAcbeta1-3GalNAcalpha beta1, 6GlcNAc-transferase, designated C2/4GnT, was identified by BLAST analysis of expressed sequence tags. The sequence of C2/4GnT encoded a putative type II transmembrane protein with significant sequence similarity to human C2GnT and IGnT. Expression of the secreted form of C2/4GnT in insect cells showed that the gene product had UDP-N-acetyl-alpha-D-glucosamine:acceptor beta1, 6-N-acetylglucosaminyltransferase (beta1,6GlcNAc-transferase) activity. Analysis of substrate specificity revealed that the enzyme catalyzed O-glycan branch formation of the core 2 and core 4 type. NMR analyses of the product formed with core 3-para-nitrophenyl confirmed the product core 4-para-nitrophenyl. The coding region of C2/4GnT was contained in a single exon and located to chromosome 15q21.3. Northern analysis revealed a restricted expression pattern of C2/4GnT mainly in colon, kidney, pancreas, and small intestine. No expression of C2/4GnT was detected in brain, heart, liver, ovary, placenta, spleen, thymus, and peripheral blood leukocytes. The expression of core 2 O-glycans has been correlated with cell differentiation processes and cancer. The results confirm the predicted existence of a beta1,6GlcNAc-transferase that functions in both core 2 and core 4 O-glycan branch formation. The redundancy in beta1,6GlcNAc-transferases capable of forming core 2 O-glycans is important for understanding the mechanisms leading to specific changes in core 2 branching during cell development and malignant transformation.