MilliporeSigma
  • Molecular cloning and characterization of a novel histone deacetylase HDAC10.
Molecular cloning and characterization of a novel histone deacetylase HDAC10.
The Journal of biological chemistry (2001-12-01)
Amaris R Guardiola, Tso-Pang Yao
ABSTRACT
The growing number of proteins controlled by reversible acetylation suggests the existence of a large number of acetyltransferases and deacetylases. Here, we report the identification of a novel class II histone deacetylase, HDAC10. Homology comparison indicates that HDAC10 is most similar to HDAC6. Both contain a unique, putative second catalytic domain not found in other HDACs. In HDAC10, however, this domain is not functional. This tandem organization of two catalytic domains confers resistance to the inhibitors trapoxin B and sodium butyrate, which potently inhibit the deacetylase activity of all other HDAC members. Thus, HDAC10 and HDAC6 share unusual structural and pharmacological characteristics. However, unlike HDAC6, which is normally a cytoplasmic deacetylase, HDAC10 resides in both the nucleus and cytoplasm. In the nucleus, when tethered to a promoter, HDAC10 represses transcription independent of its deacetylase activity, indicating that HDAC10 contains a distinct transcriptional repressor domain. These observations suggest that HDAC10 might uniquely play roles both in the nucleus, as a transcriptional modulator, and in the cytoplasm in an unidentified role. Together, our results identify HDAC10 as a novel deacetylase with distinct structure, pharmacology and localization and further expand the complexity of the HDAC family.