MilliporeSigma
  • A family of ubiquitin-like proteins binds the ATPase domain of Hsp70-like Stch.
A family of ubiquitin-like proteins binds the ATPase domain of Hsp70-like Stch.
FEBS letters (2000-02-17)
F J Kaye, S Modi, I Ivanovska, E V Koonin, K Thress, A Kubo, S Kornbluth, M D Rose
ABSTRACT
We have isolated two human ubiquitin-like (UbL) proteins that bind to a short peptide within the ATPase domain of the Hsp70-like Stch protein. Chap1 is a duplicated homologue of the yeast Dsk2 gene that is required for transit through the G2/M phase of the cell cycle and expression of the human full-length cDNA restored viability and suppressed the G2/M arrest phenotype of dsk2Delta rad23Delta Saccharomyces cerevisiae mutants. Chap2 is a homologue for Xenopus scythe which is an essential component of reaper-induced apoptosis in egg extracts. While the N-terminal UbL domains were not essential for Stch binding, Chap1/Dsk2 contains a Sti1-like repeat sequence that is required for binding to Stch and is also conserved in the Hsp70 binding proteins, Hip and p60/Sti1/Hop. These findings extend the association between Hsp70 members and genes encoding UbL sequences and suggest a broader role for the Hsp70-like ATPase family in regulating cell cycle and cell death events.