MilliporeSigma
  • The prognostic significance of ERCC1, BRCA1, XRCC1, and betaIII-tubulin expression in patients with non-small cell lung cancer treated by platinum- and taxane-based neoadjuvant chemotherapy and surgical resection.
The prognostic significance of ERCC1, BRCA1, XRCC1, and betaIII-tubulin expression in patients with non-small cell lung cancer treated by platinum- and taxane-based neoadjuvant chemotherapy and surgical resection.
Lung cancer (Amsterdam, Netherlands) (2009-08-18)
Chang Hyun Kang, Bo Gun Jang, Dong-Wan Kim, Doo Hyun Chung, Young Tae Kim, Sanghoon Jheon, Sook-Whan Sung, Joo Hyun Kim
ABSTRACT
The DNA repair pathway and isotype composition of beta-tubulin are known to be associated with resistance to platinum- and taxane-based chemotherapy, respectively. The aim of this study was to identify the clinical significance of excision repair cross-complementation 1 (ERCC1), breast cancer susceptibility gene 1 (BRCA1), X-ray repair cross-complementation 1 (XRCC1) and betaIII-tubulin on the chemotherapy response and overall survival in patients with non-small cell lung cancer (NSCLC) who received neoadjuvant chemotherapy. Protein expression profiles were evaluated by immunohistochemistry on surgical specimens of 82 NSCLC patients who underwent platinum- and taxane-based neoadjuvant chemotherapy. The expression levels of proteins were measured semi-quantitatively and the correlation with tumor responses, pathologic cell death rate and survival were evaluated. There were 73 (89.0%) clinical stage III patients. Lobectomy, bilobectomy, and pneumonectomy were performed in 54 (65.0%), 11 (13.4%), and 17 (20.7%) patients, respectively. There was no correlation between clinical response and protein expression. The expression levels of ERCC1, BRCA1, and XRCC1 increased proportionally to the cell death rate (p<0.05); however, betaIII-tubulin expression did not correlate with cell viability. Multivariate analysis demonstrated that early pathologic stage, adjuvant chemotherapy, high ERCC1 and low betaIII-tubulin expression were good prognostic factors for overall survival (p<0.05). The inverse correlation between DNA repair proteins and cell viability suggests that these protein expression levels can be markers for chemotherapy responsiveness. However, only ERCC1 and betaIII-tubulin were prognostic factors after platinum- and taxane-based neoadjuvant chemotherapy following surgical resection.