MilliporeSigma
  • L1 and epithelial cell adhesion molecules associated with gastric cancer progression and prognosis in examination of specimens from 601 patients.
L1 and epithelial cell adhesion molecules associated with gastric cancer progression and prognosis in examination of specimens from 601 patients.
Journal of experimental & clinical cancer research : CR (2014-01-16)
Yuan-Yu Wang, Li Li, Zhong-Sheng Zhao, Yong-Xiang Wang, Zai-Yuan Ye, Hou-Quan Tao
ABSTRACT
L1 cell adhesion molecule (L1CAM) and epithelial cell adhesion molecule (EPCAM) have been implicated in the development and progression of gastric cancer. The present study investigated the clinical significance of L1CAM and EPCAM in the development, progression and prognosis of gastric cancer. Expression of L1CAM and EPCAM were examined immunochemically in 601 clinicopathologically characterized gastric cancer cases. L1CAM protein was detected in 23.9% of human non-tumor mucosa samples. All samples expressed L1CAM protein at low levels. High expression of L1CAM protein was detected in 163 (27.1%) tumors. Expression of L1CAM correlated with age, tumor location, size of tumors, Lauren's classification, depth of invasion, lymph node and distant metastases, regional lymph node stage, Tumor-Node-Metastasis (TNM) stage and prognosis. EPCAM protein was detected in 45.7% of human non-tumor mucosa samples. All samples expressed EPCAM protein at low levels. High expression of EPCAM protein was detected in 247 (41.1%) tumors. Expression of EPCAM correlated with age, tumor location, size of tumors, Lauren's classification, depth of invasion, lymph node and distant metastases, regional lymph node stage, TNM stage and prognosis. Cumulative 5-year survival rates of patients with high expression of both L1CAM and EPCAM were significantly lower than in patients with low expression of both. Expression of L1CAM and EPCAM in gastric cancer was significantly associated with lymph node and distant metastasis, and poor prognosis. L1CAM and EPCAM proteins could be useful markers to predict tumor progression and prognosis.