MilliporeSigma
  • The human glutathione peroxidase genes GPX2, GPX3, and GPX4 map to chromosomes 14, 5, and 19, respectively.
The human glutathione peroxidase genes GPX2, GPX3, and GPX4 map to chromosomes 14, 5, and 19, respectively.
Cytogenetics and cell genetics (1994-01-01)
F F Chu
ABSTRACT
cDNA probes of human glutathione peroxidase (GSHPx) genes, including the classic GPX1 (GSHPx-1), the newly characterized GPX2 (GSHPx-GI), the plasma enzyme GPX3 (GSHPx-P), and the phospholipid hydroperoxide glutathione peroxidase GPX4 (PHGPX), were hybridized to Southern blots containing genomic DNA from human x hamster somatic cell hybrids. GPX2 was mapped to chromosome 14, GPX3 to chromosome 5 and GPX4 to chromosome 19. Additionally, human chromosomes 3 and 21 and the X chromosome were shown to contain sequences homologous to GPX1, as reported previously.