MilliporeSigma
  • Affinity purification probes of potential use to investigate the endogenous Hsp70 interactome in cancer.
Affinity purification probes of potential use to investigate the endogenous Hsp70 interactome in cancer.
ACS chemical biology (2014-06-18)
Anna Rodina, Tony Taldone, Yanlong Kang, Pallav D Patel, John Koren, Pengrong Yan, Erica M DaGama Gomes, Chenghua Yang, Maulik R Patel, Liza Shrestha, Stefan O Ochiana, Cristina Santarossa, Ronnie Maharaj, Alexander Gozman, Marc B Cox, Hediye Erdjument-Bromage, Ronald C Hendrickson, Leandro Cerchietti, Ari Melnick, Monica L Guzman, Gabriela Chiosis
ABSTRACT
Heat shock protein 70 (Hsp70) is a family of proteins with key roles in regulating malignancy. Cancer cells rely on Hsp70 to inhibit apoptosis, regulate senescence and autophagy, and maintain the stability of numerous onco-proteins. Despite these important biological functions in cancer, robust chemical tools that enable the analysis of the Hsp70-regulated proteome in a tumor-by-tumor manner are yet unavailable. Here we take advantage of a recently reported Hsp70 ligand to design and develop an affinity purification chemical toolset for potential use in the investigation of the endogenous Hsp70-interacting proteome in cancer. We demonstrate that these tools lock Hsp70 in complex with onco-client proteins and effectively isolate Hsp70 complexes for identification through biochemical techniques. Using these tools we provide proof-of-concept analyses that glimpse into the complex roles played by Hsp70 in maintaining a multitude of cell-specific malignancy-driving proteins.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-PCNA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution