MilliporeSigma
  • Capillary nano-immunoassay for Akt 1/2/3 and 4EBP1 phosphorylation in acute myeloid leukemia.
Capillary nano-immunoassay for Akt 1/2/3 and 4EBP1 phosphorylation in acute myeloid leukemia.
Journal of translational medicine (2014-06-14)
Himalee Sabnis, Heath L Bradley, Silvia T Bunting, Todd M Cooper, Kevin D Bunting
ABSTRACT
Overall cure rates in acute myeloid leukemia (AML) continue to range between 60-65% with disease relapse being a major cause of mortality. The PI3K-Akt-mTOR kinase pathway plays a vital role in pro-survival signals within leukemic cells and inhibition of this pathway is being investigated to improve patient outcomes. Tracking activation of multiple signaling proteins simultaneously in patient samples can be challenging especially with limiting cell numbers within rare sub-populations. The NanoPro 1000 system (ProteinSimple) is built on an automated, capillary-based immunoassay platform and enables a rapid and quantitative analysis of specific proteins and their phosphorylation states. We have utilized this nano-immunoassay to examine activation of Akt 1/2/3 and downstream mTOR target--eukaryotic initiation factor 4E-Binding Protein 1 (4EBP1). Assays for Akt 1/2/3 and 4EBP1 were standardized using AML cell lines (MV4-11, MOLM-14, OCI-AML3 and HL-60) prior to testing in patient samples. Target inhibition was studied using mTOR 1/2 inhibitor AZD-8055 and results were corroborated by Western blotting. The assay was able to quantify nanogram amounts of 4EBP1 and Akt 1/2/3 in AML cell lines and primary pediatric AML samples and results were quantifiable, consistent and reproducible. Our data provides a strong basis for testing this platform on a larger scale and our long term aim is to utilize this nano-immunoassay prospectively in de-novo AML to be able to identify poor responders who might benefit from early introduction of targeted therapy.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid