MilliporeSigma
  • Annexin A2 depletion delays EGFR endocytic trafficking via cofilin activation and enhances EGFR signaling and metastasis formation.
Annexin A2 depletion delays EGFR endocytic trafficking via cofilin activation and enhances EGFR signaling and metastasis formation.
Oncogene (2013-06-25)
M de Graauw, L Cao, L Winkel, M H A M van Miltenburg, S E le Dévédec, M Klop, K Yan, C Pont, V-M Rogkoti, A Tijsma, A Chaudhuri, R Lalai, L Price, F Verbeek, B van de Water
ABSTRACT
Enhanced epidermal growth factor receptor (EGFR) activity has been strongly linked to breast cancer progression and mediators of EGFR endocytosis may well be involved. We developed a semi-automated high-content fluorescence microscopy-based EGFR endocytosis screen to identify proteins that mediate EGFR endocytosis in human HBL100 breast cancer cells. Knockdown of 172 individual endocytosis and actin-regulatory genes with small interfering RNAs led to the identification of 14 genes of which the contribution to EGFR endocytosis in breast cancer is until now poorly defined, including DNAJC6, GDI2, FGD6, HAX1, NECAP2 and AnxA2. We show that depletion of the actin and endocytosis regulatory protein annexin A2 (AnxA2) in a panel of four triple negative breast cancer (TNBC) cell lines affected EGFR endocytosis. Depletion of AnxA2 in the aggressive and highly metastatic MDA-MB-231 TNBC cell line resulted in the inhibition of EGFR transport beyond the early endosomes. This inhibition coincided with enhanced epidermal growth factor (EGF)-induced cell migration and downstream signaling via c-Jun N-terminal kinase (JNK) and Akt. Moreover, AnxA2 knockdown increased lung metastasis formation in mice. The effect of AnxA2 knockdown on EGFR endocytosis in MDA-MB-231 was related to dephosphorylation/activation of the actin-severing protein cofilin, as re-expression of an inactive S3E-cofilin mutant, but not an active S3A-cofilin mutant, re-established EGFR endocytosis to control levels. Together, our data provide evidence for AnxA2 as a mediator of EGFR endocytosis and signaling in breast cancer via regulation of cofilin activation.