- Connecting μ-fluidics to electron microscopy.
Connecting μ-fluidics to electron microscopy.
Journal of structural biology (2011-11-19)
Simon Kemmerling, Jörg Ziegler, Gabriel Schweighauser, Stefan A Arnold, Dominic Giss, Shirley A Müller, Philippe Ringler, Kenneth N Goldie, Nils Goedecke, Andreas Hierlemann, Henning Stahlberg, Andreas Engel, Thomas Braun
PMID22094535
ABSTRACT
A versatile methodology for electron microscopy (EM) grid preparation enabling total content sample analysis is presented. A microfluidic-dialysis conditioning module to desalt or mix samples with negative stain solution is used, combined with a robotic writing table to micro-pattern the EM grids. The method allows heterogeneous samples of minute volumes to be processed at physiological pH for structure and mass analysis, and allows the preparation characteristics to be finely tuned.
Sigma-Aldrich
RPMI 1640 Vitamins Solution (100×), liquid, sterile-filtered, BioReagent, suitable for cell culture